3-Chloro-5-fluoro-4'-methyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

58819-500MG 58819-1G 58819-5G 58819-25G
77330-376EA 146.26 USD
77330-376 77345-172 77366-610 77388-320
3-Chloro-5-fluoro-4'-methyl-1,1'-biphenyl ≥97%
3-Chloro-5-fluoro-4'-methyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₁₀ClF
MW: 220.67 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758857
CAS Number: 2807450-80-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR