3'-Chloro-5'-fluoro-[1,1'-biphenyl]-3-carbaldehyde ≥95%

Supplier: AOBChem USA

Warning

58817-500MG 58817-1G 58817-5G 58817-25G
77330-374EA 188.49 USD
77330-374 77354-498 77366-608 77388-318
3'-Chloro-5'-fluoro-[1,1'-biphenyl]-3-carbaldehyde ≥95%
3'-Chloro-5'-fluoro-[1,1'-biphenyl]-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₈ClFO
MW: 234.653 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16817634
CAS Number: 1242339-78-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR