5-Fluoro-4-hydroxy-2-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

58810-500MG 58810-1G 58810-5G 58810-25G
77330-372EA 208.06 USD
77330-372 77354-494 77366-606 77388-316
5-Fluoro-4-hydroxy-2-methylbenzoic acid ≥97%
5-Fluoro-4-hydroxy-2-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₇FO₃
MW: 170.138 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22414684
CAS Number: 150164-55-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR