5-Fluoro-4-hydroxy-2-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

58807-500MG 58807-1G 58807-25G 58807-5G
77330-370EA 285.31 USD
77330-370 77354-492 77388-314 77378-600
5-Fluoro-4-hydroxy-2-methylbenzaldehyde ≥97%
5-Fluoro-4-hydroxy-2-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₇FO₂
MW: 154.138 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22414659
CAS Number: 1234845-65-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR