2-Fluoro-4-iodo-5-methylphenol ≥97%

Supplier: AOBChem USA

Warning

58805-500MG 58805-1G 58805-25G 58805-5G
77330-368EA 301.79 USD
77330-368 77354-490 77388-312 77378-598
2-Fluoro-4-iodo-5-methylphenol ≥97%
2-Fluoro-4-iodo-5-methylphenol

Precaución: for Research Use Only

Formula: C₇H₆FIO
MW: 252.025 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34551688
CAS Number: 2385268-90-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR