5-Fluoro-4-isopropoxy-2-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

58795-500MG 58795-1G 58795-25G 58795-5G
77330-366EA 301.79 USD
77330-366 77354-488 77388-310 77378-596
5-Fluoro-4-isopropoxy-2-methylbenzaldehyde ≥97%
5-Fluoro-4-isopropoxy-2-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₃FO₂
Storage Temperature: Ambient
MDL Number: MFCD34758842
CAS Number: 2807470-04-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR