4-Ethoxy-5-fluoro-2-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

58785-500MG 58785-1G 58785-5G
77337-294EA 410.97 USD
77337-294 77354-476 77378-586
4-Ethoxy-5-fluoro-2-methylbenzoic acid ≥97%
4-Ethoxy-5-fluoro-2-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁FO₃
MW: 198.191 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564584
CAS Number: 2092867-49-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR