3',5'-Dibromo-4'-methyl-[1,1'-biphenyl]-4-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

58784-500MG 58784-1G 58784-5G
77330-360EA 307.97 USD
77330-360 77354-474 77378-584
3',5'-Dibromo-4'-methyl-[1,1'-biphenyl]-4-carbaldehyde ≥97%
3',5'-Dibromo-4'-methyl-[1,1'-biphenyl]-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₁₀Br₂O
MW: 354.037 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758833
CAS Number: 2755721-63-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR