3,5-Dibromo-4'-fluoro-4-methyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

58782-500MG 58782-1G 58782-5G
77335-434EA 353.29 USD
77335-434 77354-472 77378-582
3,5-Dibromo-4'-fluoro-4-methyl-1,1'-biphenyl ≥97%
3,5-Dibromo-4'-fluoro-4-methyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₉Br₂F
MW: 344.017 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758832
CAS Number: 2755724-68-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR