3,5-Dibromo-4,4'-dimethyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

58781-500MG 58781-1G 58781-5G
77335-432EA 353.29 USD
77335-432 77354-470 77378-580
3,5-Dibromo-4,4'-dimethyl-1,1'-biphenyl ≥97%
3,5-Dibromo-4,4'-dimethyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₂Br₂
Storage Temperature: Ambient
MDL Number: MFCD34758831
CAS Number: 1236348-88-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR