3,5-Dibromo-4-methyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

58780-500MG 58780-1G 58780-5G
77335-430EA 353.29 USD
77335-430 77354-468 77378-578
3,5-Dibromo-4-methyl-1,1'-biphenyl ≥97%
3,5-Dibromo-4-methyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₁₀Br₂
MW: 326.027 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758830
CAS Number: 2755718-39-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR