2,2-Dichloro-1-(2,4-dimethoxyphenyl)ethanol ≥95%

Supplier: AOBChem USA

Warning

58762-500MG 58762-1G 58762-5G 58762-25G
77330-358EA 147.29 USD
77330-358 77345-216 77366-602 77388-306
2,2-Dichloro-1-(2,4-dimethoxyphenyl)ethanol ≥95%
2,2-Dichloro-1-(2,4-dimethoxyphenyl)ethanol

Precaución: for Research Use Only

Formula: C₁₀H₁₂Cl₂O₃
MW: 251.106 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758815
CAS Number: 2755724-67-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR