2-Bromo-6-(tert-butyl)-4-fluorophenol ≥97%

Supplier: AOBChem USA

Warning

58728-500MG 58728-1G 58728-5G 58728-25G
77330-356EA 234.84 USD
77330-356 77354-466 77366-600 77388-304
2-Bromo-6-(tert-butyl)-4-fluorophenol ≥97%
2-Bromo-6-(tert-butyl)-4-fluorophenol

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrFO
MW: 247.104 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758797
CAS Number: 2625488-77-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR