1-(Benzyloxy)-2-(tert-butyl)-4-fluorobenzene ≥95%

Supplier: AOBChem USA

Warning

58726-500MG 58726-1G 58726-5G 58726-25G
77330-354EA 264.71 USD
77330-354 77354-464 77366-598 77388-302
1-(Benzyloxy)-2-(tert-butyl)-4-fluorobenzene ≥95%
1-(Benzyloxy)-2-(tert-butyl)-4-fluorobenzene

Precaución: for Research Use Only

Formula: C₁₇H₁₉FO
Storage Temperature: Ambient
MDL Number: MFCD34758796
CAS Number: 2807450-46-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR