2-(tert-Butyl)-4-fluoro-1-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

58724-500MG 58724-1G 58724-5G
77339-784EA 638.6 USD
77339-784 77354-460 77378-574
2-(tert-Butyl)-4-fluoro-1-methoxybenzene ≥97%
2-(tert-Butyl)-4-fluoro-1-methoxybenzene

Precaución: for Research Use Only

Formula: C₁₁H₁₅FO
MW: 182.235 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29091937
CAS Number: 1598-18-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR