2,4-Dibromo-6-fluoro-N-methoxybenzamide ≥97%

Supplier: AOBChem USA

Warning

58723-500MG 58723-1G 58723-5G 58723-25G
77330-350EA 253.38 USD
77330-350 77354-458 77366-596 77388-300
2,4-Dibromo-6-fluoro-N-methoxybenzamide ≥97%
2,4-Dibromo-6-fluoro-N-methoxybenzamide

Precaución: for Research Use Only

Formula: C₈H₆Br₂FNO₂
MW: 326.945 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758794
CAS Number: 2755723-40-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR