2,4-Dibromo-6-fluoro-N-methoxy-N-methylbenzamide ≥95%

Supplier: AOBChem USA

Warning

58722-500MG 58722-1G 58722-25G 58722-5G
77330-348EA 296.64 USD
77330-348 77354-456 77388-298 77378-572
2,4-Dibromo-6-fluoro-N-methoxy-N-methylbenzamide ≥95%
2,4-Dibromo-6-fluoro-N-methoxy-N-methylbenzamide

Precaución: for Research Use Only

Formula: C₉H₈Br₂FNO₂
Storage Temperature: Ambient
MDL Number: MFCD34758793
CAS Number: 2755718-32-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR