4-(4-Bromo-3,5-difluorophenyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

58721-500MG 58721-1G 58721-5G
77340-290EA 881.68 USD
77340-290 77354-454 77378-570
4-(4-Bromo-3,5-difluorophenyl)pyridine ≥97%
4-(4-Bromo-3,5-difluorophenyl)pyridine

Precaución: for Research Use Only

Formula: C₁₁H₆BrF₂N
MW: 270.073 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758792
CAS Number: 2755723-45-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR