4'-Bromo-3',5'-difluoro-[1,1'-biphenyl]-4-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

58720-500MG 58720-1G 58720-5G 58720-25G
77330-346EA 260.59 USD
77330-346 77354-452 77366-594 77388-296
4'-Bromo-3',5'-difluoro-[1,1'-biphenyl]-4-carbaldehyde ≥97%
4'-Bromo-3',5'-difluoro-[1,1'-biphenyl]-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₇BrF₂O
MW: 297.095 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758791
CAS Number: 2755721-06-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR