4-Bromo-3,4',5-trifluoro-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

58718-500MG 58718-1G 58718-5G 58718-25G
77330-342EA 176.13 USD
77330-342 77346-558 77366-590 77388-292
4-Bromo-3,4',5-trifluoro-1,1'-biphenyl ≥97%
4-Bromo-3,4',5-trifluoro-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₂H₆BrF₃
MW: 287.075 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758789
CAS Number: 2755724-64-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR