4-Bromo-3,5-difluoro-4'-methyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

58716-500MG 58716-1G 58716-5G 58716-25G
77330-340EA 175.1 USD
77330-340 77346-486 77366-588 77388-290
4-Bromo-3,5-difluoro-4'-methyl-1,1'-biphenyl ≥97%
4-Bromo-3,5-difluoro-4'-methyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₉BrF₂
Storage Temperature: Ambient
MDL Number: MFCD34758788
CAS Number: 2764732-09-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR