2,3-Dichloro-6-iodobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

58712-500MG 58712-1G 58712-5G 58712-25G
77330-336EA 175.1 USD
77330-336 77346-484 77366-584 77388-286
2,3-Dichloro-6-iodobenzoic acid ≥97%
2,3-Dichloro-6-iodobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃Cl₂IO₂
Storage Temperature: Ambient
MDL Number: MFCD34664250
CAS Number: 219841-88-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR