1-Cyclopropyl-3,5-difluorobenzene ≥97%

Supplier: AOBChem USA

Warning

58704-500MG 58704-1G 58704-5G
77330-332EA 323.42 USD
77330-332 77354-448 77378-566
1-Cyclopropyl-3,5-difluorobenzene ≥97%
1-Cyclopropyl-3,5-difluorobenzene

Precaución: for Research Use Only

Formula: C₉H₈F₂
Storage Temperature: Ambient
MDL Number: MFCD32632587
CAS Number: 2744342-65-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR