4-(3,5-Difluorophenyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

58703-500MG 58703-1G 58703-5G
77337-672EA 435.69 USD
77337-672 77354-446 77378-564
4-(3,5-Difluorophenyl)pyridine ≥97%
4-(3,5-Difluorophenyl)pyridine

Precaución: for Research Use Only

Formula: C₁₁H₇F₂N
MW: 191.177 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15476740
CAS Number: 1166397-62-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR