3,4',5-Trifluoro-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

58700-500MG 58700-1G 58700-5G 58700-25G
77330-330EA 103 USD
77330-330 77343-036 77366-580 77378-562
3,4',5-Trifluoro-1,1'-biphenyl ≥97%
3,4',5-Trifluoro-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₂H₇F₃
MW: 208.179 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758784
CAS Number: 1555858-07-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR