3,5-Difluoro-[1,1'-biphenyl]-4-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

58695-500MG 58695-1G 58695-5G
77338-522EA 494.4 USD
77338-522 77354-442 77378-558
3,5-Difluoro-[1,1'-biphenyl]-4-carboxylic acid ≥97%
3,5-Difluoro-[1,1'-biphenyl]-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₃H₈F₂O₂
MW: 234.198 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14701763
CAS Number: 505083-02-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR