3,5-Difluoro-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

58692-500MG 58692-1G 58692-5G
77338-770EA 517.06 USD
77338-770 77354-438 77378-554
3,5-Difluoro-1,1'-biphenyl ≥97%
3,5-Difluoro-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₂H₈F₂
MW: 190.189 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14701762
CAS Number: 62351-48-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR