3',5'-Difluoro-4'-methyl-[1,1'-biphenyl]-4-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

58690-500MG 58690-1G 58690-5G 58690-25G
77330-328EA 169.95 USD
77330-328 77346-350 77366-578 77388-284
3',5'-Difluoro-4'-methyl-[1,1'-biphenyl]-4-carbaldehyde ≥97%
3',5'-Difluoro-4'-methyl-[1,1'-biphenyl]-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₁₀F₂O
MW: 232.225 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758779
CAS Number: 2764733-38-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR