3,4',5-Trifluoro-4-methyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

58688-500MG 58688-1G 58688-5G 58688-25G
77330-326EA 182.31 USD
77330-326 77346-974 77366-576 77388-282
3,4',5-Trifluoro-4-methyl-1,1'-biphenyl ≥97%
3,4',5-Trifluoro-4-methyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₉F₃
MW: 222.206 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758778
CAS Number: 2764731-62-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR