4-(Benzyloxy)-5-fluoro-2-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

58667-500MG 58667-1G 58667-5G
77337-458EA 414.06 USD
77337-458 77354-426 77378-542
4-(Benzyloxy)-5-fluoro-2-methylbenzoic acid ≥97%
4-(Benzyloxy)-5-fluoro-2-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₃FO₃
Storage Temperature: Ambient
MDL Number: MFCD34564570
CAS Number: 2734777-72-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR