Methyl 2-fluoro-4,5-dimethylbenzoate ≥97%

Supplier: AOBChem USA

Warning

58664-500MG 58664-1G 58664-5G
77339-236EA 569.59 USD
77339-236 77354-424 77378-540
Methyl 2-fluoro-4,5-dimethylbenzoate ≥97%
Methyl 2-fluoro-4,5-dimethylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁FO₂
Storage Temperature: Ambient
MDL Number: MFCD28794306
CAS Number: 1803769-81-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR