Ethyl 2-fluoro-4,5-dimethylbenzoate ≥97%

Supplier: AOBChem USA

Warning

58663-500MG 58663-1G 58663-5G
77339-234EA 569.59 USD
77339-234 77354-422 77378-538
Ethyl 2-fluoro-4,5-dimethylbenzoate ≥97%
Ethyl 2-fluoro-4,5-dimethylbenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₃FO₂
MW: 196.218 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28794292
CAS Number: 1803852-40-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR