2-Fluoro-4,5-dimethylphenol ≥95%

Supplier: AOBChem USA

Warning

58662-500MG 58662-1G 58662-5G
77339-870EA 666.41 USD
77339-870 77354-420 77378-536
2-Fluoro-4,5-dimethylphenol ≥95%
2-Fluoro-4,5-dimethylphenol

Precaución: for Research Use Only

Formula: C₈H₉FO
MW: 140.155 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15527212
CAS Number: 709031-97-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR