(2-Fluoro-4,5-dimethylphenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

58661-500MG 58661-1G 58661-5G
77338-398EA 485.13 USD
77338-398 77354-418 77378-534
(2-Fluoro-4,5-dimethylphenyl)methanol ≥95%
(2-Fluoro-4,5-dimethylphenyl)methanol

Precaución: for Research Use Only

Formula: C₉H₁₁FO
MW: 154.181 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28551038
CAS Number: 1783707-52-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR