(2-Fluoro-4,5-dimethylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

58660-500MG 58660-1G 58660-5G
77335-572EA 357.41 USD
77335-572 77354-416 77378-532
(2-Fluoro-4,5-dimethylphenyl)(methyl)sulfane ≥97%
(2-Fluoro-4,5-dimethylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₉H₁₁FS
MW: 170.247 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28794640
CAS Number: 1803819-32-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR