Methyl 3-chloro-2,5-dimethylbenzoate ≥97%

Supplier: AOBChem USA

Warning

58646-500MG 58646-1G 58646-5G
77338-046EA 460.41 USD
77338-046 77354-412 77378-526
Methyl 3-chloro-2,5-dimethylbenzoate ≥97%
Methyl 3-chloro-2,5-dimethylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁ClO₂
Storage Temperature: Ambient
MDL Number: MFCD34758756
CAS Number: 2119365-77-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR