3-Chloro-2,5-dimethylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

58643-500MG 58643-1G 58643-5G
77339-926EA 677.74 USD
77339-926 77354-408 77378-522
3-Chloro-2,5-dimethylbenzaldehyde ≥97%
3-Chloro-2,5-dimethylbenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉ClO
MW: 168.62 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28553008
CAS Number: 1784344-25-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR