1-Chloro-3-iodo-2,5-dimethylbenzene ≥97%

Supplier: AOBChem USA

Warning

58642-500MG 58642-1G 58642-5G
77337-724EA 438.78 USD
77337-724 77354-406 77378-520
1-Chloro-3-iodo-2,5-dimethylbenzene ≥97%
1-Chloro-3-iodo-2,5-dimethylbenzene

Precaución: for Research Use Only

Formula: C₈H₈ClI
MW: 266.507 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564569
CAS Number: 2382968-42-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR