4'-Bromo-2'-(trifluoromethyl)-[1,1'-biphenyl]-4-carbaldehyde ≥97%
Supplier: AOBChem USA
|
Warning
|
58632-500MG
58632-1G
58632-5G
58632-25G
77330-322EA
140.08
USD
77330-322
77344-760
77366-572
77388-280
4'-Bromo-2'-(trifluoromethyl)-[1,1'-biphenyl]-4-carbaldehyde ≥97%
4'-Bromo-2'-(trifluoromethyl)-[1,1'-biphenyl]-4-carbaldehyde
Precaución: for Research Use Only
Formula:
C₁₄H₈BrF₃O MW: 329.112 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34642019 CAS Number: 2755723-31-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...