4-Bromo-4'-fluoro-2-(trifluoromethyl)-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

58630-500MG 58630-1G 58630-5G 58630-25G
77330-320EA 122.57 USD
77330-320 77343-948 77366-570 77378-518
4-Bromo-4'-fluoro-2-(trifluoromethyl)-1,1'-biphenyl ≥97%
4-Bromo-4'-fluoro-2-(trifluoromethyl)-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₇BrF₄
MW: 319.092 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758748
CAS Number: 2755723-38-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR