4-Bromo-4'-methyl-2-(trifluoromethyl)-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

58629-500MG 58629-1G 58629-5G 58629-25G
77330-318EA 187.46 USD
77330-318 77354-404 77366-568 77388-278
4-Bromo-4'-methyl-2-(trifluoromethyl)-1,1'-biphenyl ≥97%
4-Bromo-4'-methyl-2-(trifluoromethyl)-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₀BrF₃
MW: 315.128 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758747
CAS Number: 2755719-14-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR