(2-(Trifluoromethyl)-[1,1'-biphenyl]-4-yl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

58627-500MG 58627-1G 58627-5G 58627-25G
77330-316EA 182.31 USD
77330-316 77346-972 77366-566 77388-276
(2-(Trifluoromethyl)-[1,1'-biphenyl]-4-yl)boronic acid ≥97%
(2-(Trifluoromethyl)-[1,1'-biphenyl]-4-yl)boronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₀BF₃O₂
MW: 266.024 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28167311
CAS Number: 1200834-19-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR