2-(Benzyloxy)-5-iodo-3-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

58624-500MG 58624-1G 58624-5G
77339-980EA 700.4 USD
77339-980 77354-402 77378-516
2-(Benzyloxy)-5-iodo-3-methoxybenzaldehyde ≥97%
2-(Benzyloxy)-5-iodo-3-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₅H₁₃IO₃
MW: 368.166 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758744
CAS Number: 942035-66-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR