4-(Benzyloxy)-3-formyl-5-methoxybenzoic acid ≥95%

Supplier: AOBChem USA

Warning

58622-500MG 58622-1G 58622-5G
77336-500EA 388.31 USD
77336-500 77354-400 77378-514
4-(Benzyloxy)-3-formyl-5-methoxybenzoic acid ≥95%
4-(Benzyloxy)-3-formyl-5-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₆H₁₄O₅
MW: 286.279 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758742
CAS Number: 2807467-75-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR