5-Iodo-2-isopropoxy-3-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

58616-500MG 58616-1G
77340-518EA 1421.4 USD
77340-518 77354-394
5-Iodo-2-isopropoxy-3-methoxybenzaldehyde ≥97%
5-Iodo-2-isopropoxy-3-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₃IO₃
MW: 320.124 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28158260
CAS Number: 942035-64-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR