(5-Bromo-2-isopropoxy-3-methoxyphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

58610-500MG 58610-1G 58610-5G
77330-312EA 334.75 USD
77330-312 77354-390 77378-508
(5-Bromo-2-isopropoxy-3-methoxyphenyl)methanol ≥97%
(5-Bromo-2-isopropoxy-3-methoxyphenyl)methanol

Precaución: for Research Use Only

Formula: C₁₁H₁₅BrO₃
MW: 275.139 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758731
CAS Number: 1784269-79-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR