2-Ethoxy-5-iodo-3-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

58608-500MG 58608-1G
77340-550EA 1766.45 USD
77340-550 77354-388
2-Ethoxy-5-iodo-3-methoxybenzaldehyde ≥97%
2-Ethoxy-5-iodo-3-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₁IO₃
MW: 306.097 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758729
CAS Number: 708986-45-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR