(5-Bromo-2-ethoxy-3-methoxyphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

58602-500MG 58602-1G 58602-5G
77338-382EA 484.1 USD
77338-382 77354-384 77378-504
(5-Bromo-2-ethoxy-3-methoxyphenyl)methanol ≥97%
(5-Bromo-2-ethoxy-3-methoxyphenyl)methanol

Precaución: for Research Use Only

Formula: C₁₀H₁₃BrO₃
MW: 261.112 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758723
CAS Number: 2386493-86-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR