Ethyl 3-(4-bromo-3-methoxyphenyl)-3-hydroxypropanoate ≥95%

Supplier: AOBChem USA

Warning

58588-1G 58588-5G 58588-25G
77340-756EA 60.77 USD
77340-756 77366-558 77378-500
Ethyl 3-(4-bromo-3-methoxyphenyl)-3-hydroxypropanoate ≥95%
Ethyl 3-(4-bromo-3-methoxyphenyl)-3-hydroxypropanoate

Precaución: for Research Use Only

Formula: C₁₂H₁₅BrO₄
MW: 303.149 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758718
CAS Number: 2901106-50-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR