2,4-Dibromo-N-cyclopropyl-5-fluorobenzamide ≥97%

Supplier: AOBChem USA

Warning

58538-500MG 58538-1G 58538-5G
77339-168EA 556.2 USD
77339-168 77354-374 77378-494
2,4-Dibromo-N-cyclopropyl-5-fluorobenzamide ≥97%
2,4-Dibromo-N-cyclopropyl-5-fluorobenzamide

Precaución: for Research Use Only

Formula: C₁₀H₈Br₂FNO
MW: 336.983 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758690
CAS Number: 2901096-55-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR